Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cinchonidine | 485-71-2 | 99.6% | 294.39 | 10 G
SDP

Catalog No. 5000427597
Encompass_Preferred

Cinchonidine (α-Quinidine) is a cinchona alkaloid found in *Cinchona officinalis* and *Gongronema latifolium*. It is a building block used in asymmetric synthesis in organic chemistry. It acts as a weak inhibitor of serotonin transporter (SERT) and exhibits antimalarial activities.

  • Source: Cinchona alkaloid found in *Cinchona officinalis* and *Gongronema latifolium*.
  • Application: Building block in asymmetric synthesis in organic chemistry.
  • Biological activity: Weak inhibitor of serotonin transporter (SERT).
  • Pharmacological effect: Antimalarial activities.
  • Appearance: Solid, white to off-white.
  • Storage: Room temperature for continental US shipping; 4°C, protected from light, stored under nitrogen.

Catalog No. 50-004-27597 Supplier Medchemexpress LLC Supplier No. HYN017310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cinchonidine (α-Quinidine) is a cinchona alkaloid found in *Cinchona officinalis* and *Gongronema latifolium*. It is a building block used in asymmetric synthesis in organic chemistry. It acts as a weak inhibitor of serotonin transporter (SERT) and exhibits antimalarial activities.

  • Source: Cinchona alkaloid found in *Cinchona officinalis* and *Gongronema latifolium*.
  • Application: Building block in asymmetric synthesis in organic chemistry.
  • Biological activity: Weak inhibitor of serotonin transporter (SERT).
  • Pharmacological effect: Antimalarial activities.
  • Appearance: Solid, white to off-white.
  • Storage: Room temperature for continental US shipping; 4°C, protected from light, stored under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.