Learn More
Medchemexpress LLC Cinchonidine (α-Quinidine) | 485-71-2 | 99.6% | 294.39 | 100 G
SDP
Catalog No. 5000427598
Encompass_Preferred
Cinchonidine (α-Quinidine) is a cinchona alkaloid found in *Cinchona officinalis* and *Gongronema latifolium*. It is a building block used in asymmetric synthesis in organic chemistry. It acts as a weak inhibitor of the serotonin transporter (SERT) and exhibits antimalarial activities.
  • Derived from *Cinchona officinalis* and *Gongronema latifolium*
  • Used as a building block in asymmetric synthesis
  • Weak inhibitor of the serotonin transporter (SERT)
  • Exhibits antimalarial activities
Catalog No. 50-004-27598 Supplier Medchemexpress LLC Supplier No. HYN0173100G
Cinchonidine (α-Quinidine) is a cinchona alkaloid found in *Cinchona officinalis* and *Gongronema latifolium*. It is a building block used in asymmetric synthesis in organic chemistry. It acts as a weak inhibitor of the serotonin transporter (SERT) and exhibits antimalarial activities.
  • Derived from *Cinchona officinalis* and *Gongronema latifolium*
  • Used as a building block in asymmetric synthesis
  • Weak inhibitor of the serotonin transporter (SERT)
  • Exhibits antimalarial activities