Learn More
Medchemexpress LLC Cinchonine | 118-10-5 | MFCD00064372 | 99.7% | 294.39 | C19H22N2O | 25g
SDP
Catalog No. 5000268413
Encompass_Preferred
Cinchonine is a natural compound present in Cinchona bark with antimalarial antitumor anti-inflammatory anti platelet-aggregation and anti-obesity properties Cinchonine inhibits cells proliferation and autophagy and induces apoptosis through activation of Caspase-3 Cinchonine activates endoplasmic reticulum stress-induced apoptosis in human liver cancer cells[1]
Catalog No. 50-002-68413 Supplier Medchemexpress LLC Supplier No. HYY015225G
Cinchonine is a natural compound present in Cinchona bark with antimalarial antitumor anti-inflammatory anti platelet-aggregation and anti-obesity properties Cinchonine inhibits cells proliferation and autophagy and induces apoptosis through activation of Caspase-3 Cinchonine activates endoplasmic reticulum stress-induced apoptosis in human liver cancer cells[1]