Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinchonine Hydrochloride Hydrate 80.0+%, TCI America™
SDP

Catalog No. C035125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C035125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C035125G Supplier TCI America Supplier No. C035125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5949-11-1
Molecular Formula C19H23ClN2O
Molecular Weight (g/mol) 330.86
MDL Number MFCD00079070
InChI Key IMUHWLVEEVGMBC-UHFFFAOYNA-N
PubChem CID 90472969
IUPAC Name (R)-[(2S,4R,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol;hydrochloride
SMILES C=CC1CN2CCC1CC2C(C3=CC=NC4=CC=CC=C34)O.Cl

Specifications

Melting Point 215°C
Color White
Quantity 25 g
Formula Weight 330.86
Percent Purity ≥80.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cinchonine Hydrochloride Hydrate
RTECSNumber GD2962020
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.