Learn More
Apexbio Technology LLC Cinchonine(LA40221) 118-10-5 25mg
SDP
Catalog No. 5000090681
Encompass_Preferred
Cinchonine LA40221 (118-10-5) is a small-molecule inhibitor designed to modulate biological processes such as inflammation tumorigenesis malaria infection obesity and platelet aggregation Cinchonine LA40221 exerts its biological activity primarily through regulation of signaling pathways modulation of enzyme activities and induction of apoptosis via caspase-3-dependent mechanisms In in vitro studies Cinchonine LA40221 demonstrates antitumor activity with IC50 values ranging approximately from 10-100 M depending on cell types and protocols Based on these pharmacological properties Cinchonine LA40221 holds research potential in studying malaria infection models cancer cell apoptosis inflammatory responses platelet function and obesity-related metabolic pathways
Catalog No. 50-000-90681 Supplier Apexbio Technology LLC Supplier No. C644625
Cinchonine LA40221 (118-10-5) is a small-molecule inhibitor designed to modulate biological processes such as inflammation tumorigenesis malaria infection obesity and platelet aggregation Cinchonine LA40221 exerts its biological activity primarily through regulation of signaling pathways modulation of enzyme activities and induction of apoptosis via caspase-3-dependent mechanisms In in vitro studies Cinchonine LA40221 demonstrates antitumor activity with IC50 values ranging approximately from 10-100 M depending on cell types and protocols Based on these pharmacological properties Cinchonine LA40221 holds research potential in studying malaria infection models cancer cell apoptosis inflammatory responses platelet function and obesity-related metabolic pathways