Learn More
StatLab Cincinnati Surgical Blade Handles
SDP
Catalog No. 21033339
Change view
Click to view available options
Size:
No.4 Handle
Catalog No. Size
21-033-339 No.4 Handle
1 options
Catalog No. 21-033-339 Supplier StatLab Supplier No. CSH4
Add to Cart
Add to Cart
Stainless steel and non-sterile blade handle to incorporate a satin finish, smooth loading and balance
  • Each handle is designed to incorporate a satin finish, smooth loading and balance
  • Handles are stainless steel and non-sterile
  • No. 4 handle fits No. 22 blades
  • 1 each
Specifications
Product Type Surgical Handle
Size No.4 Handle
Quantity 1 Ea.