Learn More
StatLab™ Cincinnati Surgical Blades
SDP
Catalog No. STLCSB60
Change view
Click to view available options
Size:
# 22
# 60
# 70
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
STLCSB60 # 60
STLCSB22 # 22
STLCSB70 # 70
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. STLCSB60 Supplier StatLab™ Supplier No. CSB60
Add to Cart
Add to Cart
Precision carbon blades.
  • Manufactured of high-grade surgical steel, resulting in high durability and sharpness
  • Blades are non-sterile and individually wrapped
Specifications
Product Type Surgical Blade
Size # 60
Disposable Reusable
Sterility Non-sterile
Autoclavable Not Autoclavable
Quantity 100 Blades/Pk.
Graduated No