Learn More
Cincinnati Surgical Sterile Carbon Blades
SDP
Catalog No. 19200214
Change view
Click to view available options
Size:
# 10
# 11
# 12
# 15
# 22
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-200-214 # 22
19-200-210 # 10
19-200-211 # 11
19-200-212 # 12
19-200-213 # 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-200-214 Supplier Cincinnati Surgical Supplier No. 0022
Add to Cart
Add to Cart
Packaged with VCI Paper to protect blade from corrosion
  • Made from high-grade British surgical carbon steel
  • Narrow grinding angle resulting in increased initial keenness
  • Medium surface finish
  • 200% visual inspection
  • Strong foil package
Specifications
Blade Material British Carbon Steel
Size # 22
Sterility Sterile