Learn More
Cincinnati Surgical Sterile Carbon Blades
Packaged with VCI Paper to protect blade from corrosion
$115.80 - $125.60
Specifications
Blade Material | British Carbon Steel |
---|---|
Description | Medium Surface Finish |
Sterility | Sterile |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-200-210
![]() |
Cincinnati Surgical
0010 |
# 10 |
Pack of 100 for $121.30
|
|
|||||
19-200-211
![]() |
Cincinnati Surgical
0011 |
# 11 |
Pack of 100 for $121.30
|
|
|||||
19-200-212
![]() |
Cincinnati Surgical
0012 |
# 12 |
Pack of 100 for $115.80
|
|
|||||
19-200-213
![]() |
Cincinnati Surgical
0015 |
# 15 |
Pack of 100 for $121.30
|
|
|||||
19-200-214
![]() |
Cincinnati Surgical
0022 |
# 22 |
Pack of 100 for $125.60
|
|
|||||
Description
- Made from high-grade British surgical carbon steel
- Narrow grinding angle resulting in increased initial keenness
- Medium surface finish
- 200% visual inspection
- Strong foil package
Specifications
British Carbon Steel | |
Medium Surface Finish | |
Sterile |