Learn More
Cincinnati Surgical Swann Morton Non-Sterile Carbon Steel Blades
SDP
Catalog No. 19200251
Change view
Click to view available options
Size:
# 60
Catalog No. Size
19-200-251 # 60
1 options
This item is not returnable. View return policy
Catalog No. 19-200-251 Supplier Cincinnati Surgical Supplier No. 02SM60
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use in wide variety of medical applications, including Dermaplaning
  • 200% visual inspection
  • Packaged with VCI Paper to protect blade from corrosion
  • Strongest/smallest foil package available
  • Narrow grinding angle resulting in increased initial keenness
  • High surface finish
  • Cellophane overwrap
  • Extended Range
Specifications
Blade Material Carbon Steel
Product Type Blade
Size # 60
Description High Surface Finish
Sterility Non-sterile