Learn More
Cincinnati Surgical Swann Morton Sterile Carbon Steel Blades
SDP
Catalog No. 19200229
Change view
Click to view available options
Size:
# 10
# 11
# 12
# 15
# 20
# 21
# 22
# 25A
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-200-229 # 10
19-200-230 # 11
19-200-231 # 12
19-200-232 # 15
19-200-233 # 20
19-200-234 # 21
19-200-235 # 22
19-200-236 # 25A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-200-229 Supplier Cincinnati Surgical Supplier No. 00SM10
Add to Cart
Add to Cart
For use in wide variety of medical applications, including Dermaplaning
  • 200% visual inspection
  • Packaged with VCI Paper to protect blade from corrosion
  • Strongest/smallest foil package available
  • Narrow grinding angle resulting in increased initial keenness
  • High surface finish
  • Cellophane overwrap
  • Extended Range
Specifications
Blade Material Carbon Steel
Product Type Blade
Size # 10
Description High Surface Finish
Sterility Sterile