Learn More
Cincinnati Surgical Swann Morton Sterile Stainless Steel Blades and Scalpels
SDP
Catalog No. 19200249
Change view
Click to view available options
Size:
# 10
# 10A
# 11
# 12
# 15
# 20
# 21
# 22
# 23
# 25A
Product Type:
Blade
Scalpel
Scalpel Blade
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Product Type
19-200-249 # 10A Blade
19-200-237 # 10 Blade
19-200-238 # 11 Blade
19-200-239 # 12 Blade
19-200-240 # 15 Blade
19-200-241 # 20 Blade
19-200-242 # 21 Blade
19-200-243 # 22 Blade
19-200-244 # 25A Blade
19-200-222 # 10 Scalpel
19-200-223 # 11 Scalpel Blade
19-200-224 # 15 Scalpel Blade
19-200-225 # 20 Scalpel Blade
19-200-226 # 21 Scalpel
19-200-227 # 22 Scalpel
19-200-228 # 23 Scalpel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 19-200-249 Supplier Cincinnati Surgical Supplier No. 01SM10A
Add to Cart
Add to Cart
For use in wide variety of medical applications, including Dermaplaning
  • Blades packaged with VCI Paper to protect blade from corrosion
  • Scalpels individually packaged in transparent thermos formed tray
  • 200% visual inspection
  • Strongest/Smallest foil package available
  • Narrow grinding angle resulting in increased initial keenness
  • High surface finish
  • Cellophane overwrap
  • Extended range
Specifications
Blade Material Stainless Steel
Product Type Blade
Size # 10A
Sterility Sterile
Packaging Type Packaged with VCI Paper to protect Blade from Corrosion