Learn More
Cincinnati Surgical Lance Sterile Carbon Blades
SDP
Catalog No. 19200268
Change view
Click to view available options
Size:
# 10
# 11
# 12
# 15
# 21
# 22
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-200-268 # 10
19-200-269 # 11
19-200-270 # 12
19-200-271 # 15
19-200-272 -
19-200-273 # 21
19-200-274 # 22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-200-268 Supplier Cincinnati Surgical Supplier No. 92010
Add to Cart
Add to Cart
Made from British steel, offering unique shape, size and material composition
  • 100% visual inspection
  • Wide grinding angle for high durability
  • Low surface finish
Certifications
CE Marked
Specifications
Blade Material British Cold-rolled Surgical Carbon Steel
Product Type Handle
Size # 10
Description Low Surface Finish
Sterility Sterile
Color Green