Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cineole, Approx. 99%, MP Biomedicals™

Catalog No. ICN15068950 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15068950 50 mL
ICN15068990 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15068950 Supplier MP Biomedicals, Inc Supplier No. 0215068950
Add to Cart
Add to Cart
Pharmaceuticals, Insecticide
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 470-82-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00167977
InChI Key WEEGYLXZBRQIMU-UHFFFAOYSA-N
Synonym eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan
PubChem CID 2758
IUPAC Name 1,3,3-trimethyl-2-oxabicyclo[2.2.2]octane
SMILES CC12CCC(CC1)C(C)(C)O2

Specifications

Color Colorless
Density 0.921 to 0.924g/mL
Assay Percent Range Approx. 99%
Quantity 50 mL
Formula Weight 154.3
Physical Form Liquid
Chemical Name or Material Cineole
EINECSNumber 207-431-5
Recommended Storage Store at room temperature (15° to 30°C).
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.