Learn More
Apexbio Technology LLC Cinepazide maleate 26328-04-1 50mg
SDP
Catalog No. 5000090316
Encompass_Preferred
Cinepazide maleate (CAS 26328-04-1) is the maleate salt of cinepazide classified as a vasodilatory agent It exerts its effects primarily through inhibition of calcium influx in vascular smooth muscle cells leading to vessel relaxation and increased blood flow This mechanism is associated with reduced peripheral resistance and enhanced microcirculation Cinepazide maleate has been utilized in research exploring vascular tone modulation cerebral blood flow and ischemia models providing a tool for investigating mechanisms of vasodilation and circulatory disorders in preclinical settings
Catalog No. 50-000-90316 Supplier Apexbio Technology LLC Supplier No. B191150
Cinepazide maleate (CAS 26328-04-1) is the maleate salt of cinepazide classified as a vasodilatory agent It exerts its effects primarily through inhibition of calcium influx in vascular smooth muscle cells leading to vessel relaxation and increased blood flow This mechanism is associated with reduced peripheral resistance and enhanced microcirculation Cinepazide maleate has been utilized in research exploring vascular tone modulation cerebral blood flow and ischemia models providing a tool for investigating mechanisms of vasodilation and circulatory disorders in preclinical settings