Learn More
Medchemexpress LLC Cinepazide maleate | 26328-04-1 | 99.7% | 533.57 g/mol | C26H35N3O9 | 10 MG
SDP
Catalog No. 5000298793
Encompass_Preferred
Cinepazide maleate is a piperazine-derivative vasodilator and weak calcium channel blocker used in preclinical research into cerebrovascular and cardiovascular disorders. It is supplied as a white to off-white solid of high purity and is intended for in vitro and in vivo studies focused on vascular function and ion-channel modulation.
  • Vasodilator and weak calcium channel blocker used in vascular research.
  • High purity (99.7%) suitable for reproducible experimental results.
  • White to off-white solid for standard laboratory handling.
  • Recommended sealed storage away from moisture to maintain stability.
  • Supplied in small research quantities for preclinical use.
Catalog No. 50-002-98793 Supplier Medchemexpress LLC Supplier No. HY6601010MG
Cinepazide maleate is a piperazine-derivative vasodilator and weak calcium channel blocker used in preclinical research into cerebrovascular and cardiovascular disorders. It is supplied as a white to off-white solid of high purity and is intended for in vitro and in vivo studies focused on vascular function and ion-channel modulation.
  • Vasodilator and weak calcium channel blocker used in vascular research.
  • High purity (99.7%) suitable for reproducible experimental results.
  • White to off-white solid for standard laboratory handling.
  • Recommended sealed storage away from moisture to maintain stability.
  • Supplied in small research quantities for preclinical use.