Learn More
Medchemexpress LLC Cinepazide (Maleate) | 26328-04-1 | 99.7% | C26H35N3O9 | 1 ML
SDP
Catalog No. 5000433924
Encompass_Preferred
Cinepazide Maleate is a piperazine derivative that acts as a weak calcium channel blocker and a potent vasodilator. It is used in research for cerebrovascular diseases, including ischemic stroke and brain infarct.
  • Acts as a piperazine derivative
  • Functions as a weak calcium channel blocker
  • Potent vasodilator
  • Useful in research for cerebrovascular diseases
  • Suitable for ischemic stroke studies
  • Applicable for brain infarct research
Catalog No. 50-004-33924 Supplier Medchemexpress LLC Supplier No. HY6601010MM/1ML
Cinepazide Maleate is a piperazine derivative that acts as a weak calcium channel blocker and a potent vasodilator. It is used in research for cerebrovascular diseases, including ischemic stroke and brain infarct.
  • Acts as a piperazine derivative
  • Functions as a weak calcium channel blocker
  • Potent vasodilator
  • Useful in research for cerebrovascular diseases
  • Suitable for ischemic stroke studies
  • Applicable for brain infarct research