Learn More
Medchemexpress LLC Cinepazide Maleate | 26328-04-1 | 99.7% | C26H35N3O9 | 25 MG
SDP
Catalog No. 5000359230
Encompass_Preferred
Cinepazide Maleate is a piperazine derivative that acts as a weak calcium channel blocker and a potent vasodilator. It is utilized in research for cerebrovascular diseases, including ischemic stroke and brain infarct.
  • Acts as a weak calcium channel blocker
  • Potent vasodilator
  • Used for research on cerebrovascular diseases
  • Suitable for ischemic stroke and brain infarct research
  • For research use only
Catalog No. 50-003-59230 Supplier Medchemexpress LLC Supplier No. 25MGHY66010
Cinepazide Maleate is a piperazine derivative that acts as a weak calcium channel blocker and a potent vasodilator. It is utilized in research for cerebrovascular diseases, including ischemic stroke and brain infarct.
  • Acts as a weak calcium channel blocker
  • Potent vasodilator
  • Used for research on cerebrovascular diseases
  • Suitable for ischemic stroke and brain infarct research
  • For research use only