Learn More
Medchemexpress LLC Cinepazide Maleate | 26328-04-1 | C26H35N3O9 | 100 MG
SDP
Catalog No. 5000434991
Encompass_Preferred
Cinepazide Maleate is a piperazine derivative that acts as a weak calcium channel blocker and a potent vasodilator. It is used for the research of cerebrovascular diseases, including ischemic stroke and brain infarct. The salt form offers enhanced water solubility and stability, making it suitable for various research applications. This product is for research use only and not intended for patient use.
  • Piperazine derivative
  • Weak calcium channel blocker
  • Potent vasodilator
  • Used for cerebrovascular disease research
  • Enhanced water solubility and stability
  • For research use only
Catalog No. 50-004-34991 Supplier Medchemexpress LLC Supplier No. HY66010100MG
Cinepazide Maleate is a piperazine derivative that acts as a weak calcium channel blocker and a potent vasodilator. It is used for the research of cerebrovascular diseases, including ischemic stroke and brain infarct. The salt form offers enhanced water solubility and stability, making it suitable for various research applications. This product is for research use only and not intended for patient use.
  • Piperazine derivative
  • Weak calcium channel blocker
  • Potent vasodilator
  • Used for cerebrovascular disease research
  • Enhanced water solubility and stability
  • For research use only