Learn More
Abcam Cinnabarinic acid, Endogenous mGlu4 selective agonist, 50MG
Catalog No. 502707539
Encompass_Preferred
MW 300.22 Da, Purity >98%. Endogenous selective mGlu4 agonist; generated by autoxidation of the tryptophan metabolite 3-hydroxyanthranilic acid. Activates mGlu4 receptors in transiently transfected HEK293 cells, and is devoid of activity on other mGlu receptor subtypes (mGlu1, 2, 5, 6, 7, and 8). Protects against NMDA-induced excitotoxic cell death.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7539 Supplier Abcam Supplier No. AB12050650MG
MW 300.22 Da, Purity >98%. Endogenous selective mGlu4 agonist; generated by autoxidation of the tryptophan metabolite 3-hydroxyanthranilic acid. Activates mGlu4 receptors in transiently transfected HEK293 cells, and is devoid of activity on other mGlu receptor subtypes (mGlu1, 2, 5, 6, 7, and 8). Protects against NMDA-induced excitotoxic cell death.
The product is subject to the following: Abcam Restricted Use Statement