Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnamamide, predominantly trans, 97%

Catalog No. AAA1342906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1342906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1342906 Supplier Thermo Scientific Chemicals Supplier No. A1342906

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

CAS: 621-79-4 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-79-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00008033
InChI Key APEJMQOBVMLION-VOTSOKGWSA-N
Synonym cinnamamide, trans-cinnamamide, 3-phenylacrylamide, cinnamic amide, 2-propenamide, 3-phenyl, 3-phenylpropenamide, 2-benzylideneacetamide, e-cinnamamide, ginnamic acid amide, trans-cinnamoylamine
PubChem CID 5273472
ChEBI CHEBI:76320
IUPAC Name (E)-3-phenylprop-2-enamide
SMILES C1=CC=C(C=C1)C=CC(=O)N

Specifications

Melting Point 145°C to 149°C
Quantity 5 g
Beilstein 2040577
Formula Weight 147.18
Percent Purity 97%
Chemical Name or Material Cinnamamide, predominantly trans
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 210-707-8
RTECSNumber GD6853000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.