Learn More
Selleck Chemical LLC Cinnamic acid S3677-25mg
SDP
Catalog No. 503108368
Encompass_Preferred
Cinnamic acid (Benzenepropenoic acid Isocinnamic acid trans-Cinnamic acid Phenylacrylic acid) a naturally occurring aromatic fatty acid of low toxicity induces cytostasis and a reversal of malignant properties of human tumor cells in vitro
Catalog No. 50-310-8368 Supplier Selleck Chemical LLC Supplier No. S367725MG
Cinnamic acid (Benzenepropenoic acid Isocinnamic acid trans-Cinnamic acid Phenylacrylic acid) a naturally occurring aromatic fatty acid of low toxicity induces cytostasis and a reversal of malignant properties of human tumor cells in vitro