Learn More
Apexbio Technology LLC Cinnamic acid 621-82-9 100mg
SDP
Catalog No. 5000095905
Encompass_Preferred
Cinnamic acid (CAS 621-82-9) is an aromatic unsaturated carboxylic acid naturally occurring in various plants particularly in cinnamon and vanilla It exhibits a range of bioactivities including antimicrobial anti-inflammatory and antioxidant effects Mechanistically cinnamic acid has been shown to modulate inflammatory pathways and oxidative stress responses thereby influencing cellular signaling cascades associated with disease progression Owing to these properties it is investigated in biomedical research for its potential in cancer intervention as well as for broader applications in pharmacological studies of inflammation and infection
Catalog No. 50-000-95905 Supplier Apexbio Technology LLC Supplier No. A9923100
Cinnamic acid (CAS 621-82-9) is an aromatic unsaturated carboxylic acid naturally occurring in various plants particularly in cinnamon and vanilla It exhibits a range of bioactivities including antimicrobial anti-inflammatory and antioxidant effects Mechanistically cinnamic acid has been shown to modulate inflammatory pathways and oxidative stress responses thereby influencing cellular signaling cascades associated with disease progression Owing to these properties it is investigated in biomedical research for its potential in cancer intervention as well as for broader applications in pharmacological studies of inflammation and infection