Learn More
Medchemexpress LLC Cinnamic acid-d6 | 91453-04-2 | 154.20 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0610AS5MG
Encompass_Preferred
Cinnamic acid-d6 is the deuterium labeled Cinnamic acid. Cinnamic acid has potential use in cancer intervention, with IC50s of 1-4.5 mM in glioblastoma, melanoma, prostate and lung carcinoma cells. For research use only. Not sold to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88707