Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnamic Aldehyde, 99%, MP Biomedicals

Catalog No. ICN15069080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15069080 100 g
ICN15069090 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15069080 Supplier MP Biomedicals, Inc Supplier No. 0215069080
Add to Cart
Add to Cart

Chemical Identifiers

CAS 104-55-2
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
InChI Key KJPRLNWUNMBNBZ-QPJJXVBHSA-N
Synonym cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
PubChem CID 637511
ChEBI CHEBI:16731
IUPAC Name (E)-3-phenylprop-2-enal
SMILES C1=CC=C(C=C1)C=CC=O

Specifications

Assay Percent Range 99%
Quantity 100 g
Formula Weight 132.2
Concentration or Composition (by Analyte or Components) 1mL = approx. 1.05g
Chemical Name or Material Cinnamic Aldehyde
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.