Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnamic Aldehyde, MP Biomedicals™

Catalog No. mp215069091 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215069091 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215069091 Supplier MP Biomedicals, Inc Supplier No. 0215069091
Only null left
Add to Cart
Add to Cart

Cinnamic Aldehyde, MP Biomedicals is also known as Cinnamaldehyde.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-55-2
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
InChI Key KJPRLNWUNMBNBZ-QPJJXVBHSA-N
Synonym cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
PubChem CID 637511
ChEBI CHEBI:16731
IUPAC Name (E)-3-phenylprop-2-enal
SMILES C1=CC=C(C=C1)C=CC=O

Specifications

Melting Point -7.5°C (literature)
Density 1.0497g/cm3 at 20°C (Lit.)
Boiling Point 246°C (1014 hPa (literature))
Refractive Index n20/D 1.621 (Lit.)
Quantity 1000 g
Solubility Information Misc with alcohol, ether, chloroform, oils.
Formula Weight 132.2
Percent Purity ≥99%
Physical Form Liquid
Recommended Storage Store at 4°C, under nitrogen.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.