Learn More
Medchemexpress LLC Cinnamoylglycine-2,2-d2 | 1219806-46-8 | 99.4% | C11H9D2NO3 | 5 MG
SDP
Catalog No. 5000434979
Encompass_Preferred
Cinnamoylglycine-2,2-d2 is a chemical compound with the molecular formula C11H9D2NO3 and CAS number 1219806-46-8. It exhibits a purity of 99.4% by HPLC and appears as a white to off-white solid. This product is intended for research use only.
  • Consistent with structure by 1H NMR spectrum
  • Isotopic enrichment of 98.96%
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-34979 Supplier Medchemexpress LLC Supplier No. HY77641S5MG
Cinnamoylglycine-2,2-d2 is a chemical compound with the molecular formula C11H9D2NO3 and CAS number 1219806-46-8. It exhibits a purity of 99.4% by HPLC and appears as a white to off-white solid. This product is intended for research use only.
  • Consistent with structure by 1H NMR spectrum
  • Isotopic enrichment of 98.96%
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month