Learn More
Enzo Life Sciences Cinnamtannin B-1 (5mg). CAS: 88082-60-4
SDP
Catalog No. 502008866
Encompass_Preferred
A-type proanthocyanidin contained in several plant species such as Laurus nobilis L. , Vaccinium vitis-idaea, Parameria laevigata, Cinnamomum zeylanicum, Lindera umbellata, Metaxya rostrata. Potent antioxidant, TEAC method (nM Trolox) = 0.95. Protective agent against oxidative stress and apoptosis in human platelets. Inhibitory activity against cyclooxygenase-2 (COX-2) in vitro. DPPH radical scavenging IC50 6.14µM Lipid peroxidation inhibition 159.74 Nitric oxide inhibition 10.28 Antibacterial against B.subtilis (9mm, 20mg/ml) Alternative name: Epicatechin-(4beta->8,2beta->O->7)-epicatechin-(4alpha->8)-epicatechin. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol, methanol or in a mixture of DMSO and water. Long Term Storage: -20°C.
Catalog No. 50-200-8866 Supplier Enzo Life Sciences Supplier No. ALX350365M005
Add to Cart
Add to Cart
A-type proanthocyanidin contained in several plant species such as Laurus nobilis L. , Vaccinium vitis-idaea, Parameria laevigata, Cinnamomum zeylanicum, Lindera umbellata, Metaxya rostrata. Potent antioxidant, TEAC method (nM Trolox) = 0.95. Protective agent against oxidative stress and apoptosis in human platelets. Inhibitory activity against cyclooxygenase-2 (COX-2) in vitro. DPPH radical scavenging IC50 6.14µM Lipid peroxidation inhibition 159.74 Nitric oxide inhibition 10.28 Antibacterial against B.subtilis (9mm, 20mg/ml) Alternative name: Epicatechin-(4beta->8,2beta->O->7)-epicatechin-(4alpha->8)-epicatechin. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol, methanol or in a mixture of DMSO and water. Long Term Storage: -20°C.