Learn More
Medchemexpress LLC Cinnamyl-3,4-dihydroxy-α-cyanocinnamate | 132465-11-3 | 99.41% | 321.33 | 25 MG
SDP
Catalog No. 5000350122
Encompass_Preferred
Cinnamyl-3,4-dihydroxy-α-cyanocinnamate (CDC) is a potent inhibitor of 12/15-Lipoxygenases (LO) with potential for research into type 1 diabetes mellitus. It has been shown to antagonize the effect of high glucose on protein phosphorylation to mitigate destruction of the endothelial cell barrier in a mouse diabetes mellitus model. In vitro, CDC partially suppressed dextran transport increased by high glucose or 12(S)-HETE.
  • Potent inhibitor of 12/15-Lipoxygenases (LO)
  • Potential for type 1 diabetes mellitus research
  • Antagonizes high glucose effect on protein phosphorylation
  • Mitigates endothelial cell barrier destruction in mouse models
  • Partially suppresses dextran transport increased by high glucose or 12(S)-HETE
Catalog No. 50-003-50122 Supplier Medchemexpress LLC Supplier No. 25MGHY138688
Cinnamyl-3,4-dihydroxy-α-cyanocinnamate (CDC) is a potent inhibitor of 12/15-Lipoxygenases (LO) with potential for research into type 1 diabetes mellitus. It has been shown to antagonize the effect of high glucose on protein phosphorylation to mitigate destruction of the endothelial cell barrier in a mouse diabetes mellitus model. In vitro, CDC partially suppressed dextran transport increased by high glucose or 12(S)-HETE.
  • Potent inhibitor of 12/15-Lipoxygenases (LO)
  • Potential for type 1 diabetes mellitus research
  • Antagonizes high glucose effect on protein phosphorylation
  • Mitigates endothelial cell barrier destruction in mouse models
  • Partially suppresses dextran transport increased by high glucose or 12(S)-HETE