Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cinnamyl acetate | 103-54-8 | 500 G
SDP

Catalog No. 5000419761
Encompass_Preferred

Cinnamyl acetate is a chemical compound widely utilized in the flavor and fragrance industry, and it also acts as a broad-spectrum antibacterial agent.

  • Functions as a broad-spectrum antibacterial agent
  • Used extensively in the flavor and fragrance industry
  • Molecular formula C11H12O2
  • Molecular weight 176.21
  • Appears as a colorless to light yellow liquid
  • Stable under recommended storage conditions

Catalog No. 50-004-19761 Supplier Medchemexpress LLC Supplier No. HYN7125500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cinnamyl acetate is a chemical compound widely utilized in the flavor and fragrance industry, and it also acts as a broad-spectrum antibacterial agent.

  • Functions as a broad-spectrum antibacterial agent
  • Used extensively in the flavor and fragrance industry
  • Molecular formula C11H12O2
  • Molecular weight 176.21
  • Appears as a colorless to light yellow liquid
  • Stable under recommended storage conditions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.