Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cinnamyl acetate | 103-54-8 | MFCD00008722 | 1 ML
SDP

Catalog No. 5000417592
Encompass_Preferred

Cinnamyl acetate is widely applied in the flavor and fragrance industry and functions as a new broad-spectrum antibacterial agent. It is sourced from plants like Liliaceae Allium sativum Linn.

  • Purity of 99.39%
  • Molecular weight is 176.21
  • Chemical formula is C11H12O2
  • Appears as a liquid
  • Colorless to light yellow appearance
  • Initially sourced from Liliaceae Allium sativum Linn. plants
  • Ships at room temperature within the continental US (may vary elsewhere)
  • Storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL (requires ultrasonic)
  • Solubility in vivo protocols available for clear solutions at ≥ 2.5 mg/mL

Catalog No. 50-004-17592 Supplier Medchemexpress LLC Supplier No. HYN712510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cinnamyl acetate is widely applied in the flavor and fragrance industry and functions as a new broad-spectrum antibacterial agent. It is sourced from plants like Liliaceae Allium sativum Linn.

  • Purity of 99.39%
  • Molecular weight is 176.21
  • Chemical formula is C11H12O2
  • Appears as a liquid
  • Colorless to light yellow appearance
  • Initially sourced from Liliaceae Allium sativum Linn. plants
  • Ships at room temperature within the continental US (may vary elsewhere)
  • Storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL (requires ultrasonic)
  • Solubility in vivo protocols available for clear solutions at ≥ 2.5 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.