Learn More
Medchemexpress LLC Cinnamyl alcohol | 104-54-1 | MFCD00002921 | 134.18 | C9H10O | 5g
SDP
Catalog No. 5000268560
Encompass_Preferred
Cinnamyl Alcohol is an active component from chestnut flower inhibits increased PPAR expression exhibits anti-obesity antioxidant and anti-inflammatory activities[1]
Catalog No. 50-002-68560 Supplier Medchemexpress LLC Supplier No. HYY00785G
Cinnamyl Alcohol is an active component from chestnut flower inhibits increased PPAR expression exhibits anti-obesity antioxidant and anti-inflammatory activities[1]