Learn More
Medchemexpress LLC Cinnamyl alcohol | 104-54-1 | MFCD00002921 | 99.4% | 134.18 g/mol | C9H10O | 5mg
SDP
Catalog No. 5000268938
Encompass_Preferred
Cinnamyl Alcohol (Standard) is the analytical standard of Cinnamyl Alcohole (HY-Y0078) This product is intended for research and analytical applications Cinnamyl Alcohol is an active component from chestnut flower inhibits increased PPAR expression exhibits anti-obesity antioxidant and anti-inflammatory activities[1]
Catalog No. 50-002-68938 Supplier Medchemexpress LLC Supplier No. HYY0078R5MG
Cinnamyl Alcohol (Standard) is the analytical standard of Cinnamyl Alcohole (HY-Y0078) This product is intended for research and analytical applications Cinnamyl Alcohol is an active component from chestnut flower inhibits increased PPAR expression exhibits anti-obesity antioxidant and anti-inflammatory activities[1]