Learn More
Strem, An Ascensus Company [(Cinnamyl)PdCl]2, 98% 2g
SDP
Supplier:  Strem, An Ascensus Company 4602952G
Encompass
CAS# 12131-44-1. 2g. Di-mu-chlorobis[(1,2,3-η;)-1-phenyl-2-propenyl]dipalladium(II), 98%. MFCD16621470. Molecular Weight: 518.08. Molecular Formula: [(C9H9)ClPd]2. Color/form: yellow xtl. Strem# 46-0295. http://www.strem.com/catalog/v/46-0295/
Catalog No. 50-203-4151