Learn More
Medchemexpress LLC HY-N7129 100mg Medchemexpress, Cinnamylideneacetic acid CAS:1552-94-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN7129100MG
Encompass_Preferred
Medchemexpress, HY-N7129 100mg Cinnamylideneacetic acid CAS:1552-94-9 Cinnamylideneacetic acid is a photoresponsive compound which is capable of a photoinduced [2+2] cycloaddition [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3985