Learn More
AdipoGen CinnamyltriphenylphosphoniumCl
SDP
Supplier:  AdipoGen CDXC0156G100
Encompass_Preferred
Chemical. CAS 1530-35-4. Formula C27H24ClP. MW 414.91. Synthetic Building block for synthesis. Used in Wittig reactions.
Catalog No. 50-256-1449