Learn More
Medchemexpress LLC Cinnarizine | 298-57-7 | MFCD00056037 | 99.0% | 25 MG
SDP
Catalog No. 5000413875
Encompass_Preferred
Cinnarizine (Standard) is an analytical standard. It is an orally active, effective and selective inhibitor of L-type calcium channel Cav1.3. This product is intended for research and analytical applications.
  • Inhibits L-type calcium channel Cav1.3 with an IC50 of 1.5 μM.
  • Crosses the blood-brain barrier to regulate calcium homeostasis and dopamine neurotransmission.
  • Inhibits influx of calcium ions into smooth muscle cells, relaxing vascular smooth muscle.
  • Improves cerebral circulation and reduces blood viscosity.
  • Antagonizes dopamine receptors.
  • Used in the study of vestibular vertigo, Meniere's disease, and cerebrovascular diseases.
Catalog No. 50-004-13875 Supplier Medchemexpress LLC Supplier No. 25MGHYB1090R
Cinnarizine (Standard) is an analytical standard. It is an orally active, effective and selective inhibitor of L-type calcium channel Cav1.3. This product is intended for research and analytical applications.
  • Inhibits L-type calcium channel Cav1.3 with an IC50 of 1.5 μM.
  • Crosses the blood-brain barrier to regulate calcium homeostasis and dopamine neurotransmission.
  • Inhibits influx of calcium ions into smooth muscle cells, relaxing vascular smooth muscle.
  • Improves cerebral circulation and reduces blood viscosity.
  • Antagonizes dopamine receptors.
  • Used in the study of vestibular vertigo, Meniere's disease, and cerebrovascular diseases.