Learn More
Enzo Life Sciences CinnGEL 2-methylester (1mg)
SDP
Supplier:  Enzo Life Sciences BMLPR1150001
Encompass_Preferred
Novel peptoid inhibitor of PTP1B (IC50=1.3µM). Methyl ester (for cell permeability) is hydrolyzed to active inhibitor by intracellular esterases. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1512