Learn More
Apexbio Technology LLC Cinobufagin 470-37-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062821
Encompass_Preferred
Cinobufagin (CAS 470-37-1) is a steroidal cardiac glycoside recognized for its inhibitory effect on the Na /K -ATPase pump Its inhibitory potency is approximately comparable to that of ouabain By impeding Na /K -ATPase activity cinobufagin disrupts ionic gradients altering intracellular calcium homeostasis and influencing cardiac and cellular functions In biomedical research cinobufagin serves as a valuable pharmacological tool to investigate ion transport mechanisms and explore related signaling pathways implicated in cellular physiology and pathology
Catalog No. 50-000-62821 Supplier Apexbio Technology LLC Supplier No. N11545.1
Cinobufagin (CAS 470-37-1) is a steroidal cardiac glycoside recognized for its inhibitory effect on the Na /K -ATPase pump Its inhibitory potency is approximately comparable to that of ouabain By impeding Na /K -ATPase activity cinobufagin disrupts ionic gradients altering intracellular calcium homeostasis and influencing cardiac and cellular functions In biomedical research cinobufagin serves as a valuable pharmacological tool to investigate ion transport mechanisms and explore related signaling pathways implicated in cellular physiology and pathology