Learn More
Apexbio Technology LLC Cinobufagin, 20mg. Cas: 470-37-1 MFCD: MFCD00056825.
SDP
Catalog No. 501152823
Encompass_Preferred
Cinobufagin is a specific Na+/K+-ATPase inhibitor. About as active as ouabain. Other sizes are available. Please inqury us for quote.
Catalog No. 50-115-2823 Supplier Apexbio Technology LLC Supplier No. N115420
Cinobufagin is a specific Na+/K+-ATPase inhibitor. About as active as ouabain. Other sizes are available. Please inqury us for quote.