Learn More
Biomatik Corporation Cinobufagin (CBG) ELISA Kit
Supplier:  Biomatik Corporation EKU10719
Encompass_Preferred
Cinobufagin (CBG) ELISA Kit for detecting CBG in biological agents, other biological fluids. Species Reactivity: Universal. Detection Range: 246.9-20,000ng/ml. Sensitivity: 91.3ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7027