Learn More
TARGETMOL CHEMICALS INC CINOBUFOTALIN 10MG
SDP
Catalog No. 502579841
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cinobufotalin is a main cardiac toxin in toad it is a novel anti-HCC agent it induces growth inhibition and apoptosis in cultured HCC cells through ceramide production. Cinobufotalin is also an effective reversal agent for the multidrug resistance of tumors it can reverse the adriamycin-resistance in Raji/ADR cells and the expression of P-gp and MRP-1 proteins. Cinobufotalin can promote the dendritic cells(DCs) derived from peripheral blood of patients with chronic hepatitis B to mature and effectively enhance its(the DCs') capabilities therefore the treatment of cinobufotalin may potentiate the antiviral immunity of the patients with chronic hepatitis B(CHB). purity: 99%
Catalog No. 50-257-9841 Supplier TARGETMOL CHEMICALS INC Supplier No. T4A239910MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cinobufotalin is a main cardiac toxin in toad it is a novel anti-HCC agent it induces growth inhibition and apoptosis in cultured HCC cells through ceramide production. Cinobufotalin is also an effective reversal agent for the multidrug resistance of tumors it can reverse the adriamycin-resistance in Raji/ADR cells and the expression of P-gp and MRP-1 proteins. Cinobufotalin can promote the dendritic cells(DCs) derived from peripheral blood of patients with chronic hepatitis B to mature and effectively enhance its(the DCs') capabilities therefore the treatment of cinobufotalin may potentiate the antiviral immunity of the patients with chronic hepatitis B(CHB). purity: 99%