Learn More
Apexbio Technology LLC CINOXACIN-200MG
SDP
Catalog No. 5000569136
Encompass_Preferred
Cinoxacin (CAS No 28657-80-9) is an antibacterial agent belonging to the quinolone class of antibiotics Originally developed for treating urinary tract infections it functions by inhibiting bacterial DNA gyrase thereby preventing bacterial DNA replication and transcription In vitro studies demonstrate activity in the micromolar range against various gram-negative bacteria particularly effective in inhibiting Escherichia coli Cinoxacin is frequently used in preclinical research involving cell cultures and animal models to evaluate its antimicrobial efficacy and pharmacokinetics It serves as a tool in drug screening to explore novel quinolone derivatives Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69136 Supplier Apexbio Technology LLC Supplier No. BA1045200
Cinoxacin (CAS No 28657-80-9) is an antibacterial agent belonging to the quinolone class of antibiotics Originally developed for treating urinary tract infections it functions by inhibiting bacterial DNA gyrase thereby preventing bacterial DNA replication and transcription In vitro studies demonstrate activity in the micromolar range against various gram-negative bacteria particularly effective in inhibiting Escherichia coli Cinoxacin is frequently used in preclinical research involving cell cultures and animal models to evaluate its antimicrobial efficacy and pharmacokinetics It serves as a tool in drug screening to explore novel quinolone derivatives Experimental concentrations typically depend on the specific research design