Learn More
Medchemexpress LLC HY-110249 10mg Medchemexpress, CINPA1 CAS:102636-74-8 Purity:>98%
SDP
Catalog No. 501968775
Encompass_Preferred
Medchemexpress, HY-110249 10mg CINPA1 CAS:102636-74-8 CINPA1 is a potent and specific inhibitor of constitutive androstane receptor (CAR) that does not activate pregnane X receptor (PXR). CINPA1 reduces CAR-mediated transcription with an IC50 of ~70 nM. CINPA1 can be used as a molecular tool for understanding CAR function. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8775 Supplier Medchemexpress LLC Supplier No. HY11024910MG
Medchemexpress, HY-110249 10mg CINPA1 CAS:102636-74-8 CINPA1 is a potent and specific inhibitor of constitutive androstane receptor (CAR) that does not activate pregnane X receptor (PXR). CINPA1 reduces CAR-mediated transcription with an IC50 of ~70 nM. CINPA1 can be used as a molecular tool for understanding CAR function. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.