Learn More
Medchemexpress LLC Cintirorgon | 2055536-64-4 | MFCD31544352 | 99.1% | 603.53 g/mol | C27H23F6NO6S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1040371MG
Encompass_Preferred
Cintirorgon is an orally bioavailable small-molecule agonist of retinoic acid-related orphan receptor gamma (RORγ), provided as a research-grade reference compound for studying RORγ-driven signaling in cellular and animal models. It is characterized by a well-defined chemical profile and high purity suitable for pharmacology and immunology applications.
  • Orally bioavailable RORγ agonist.
  • High purity suitable for analytical and biological assays.
  • Prepared for use in in vitro reporter assays and cellular studies.
  • Applicable to in vivo pharmacology and immunomodulatory investigations.
  • Soluble in standard organic solvents for formulation and dosing.
Catalog No. 50-002-43211