Learn More
Medchemexpress LLC Cintirorgon | 2055536-64-4 | MFCD31544352 | 99.1% | 603.53 g/mol | C27H23F6NO6S | 50 MG
SDP
Catalog No. 5000244514
Encompass_Preferred
Cintirorgon (LYC-55716) is a selective, orally bioavailable retinoic acid receptor-related orphan receptor γ (RORγ) agonist used as a research tool to modulate RORγ-dependent gene expression in T lymphocytes. It is provided as a high-purity solid suitable for in vitro and in vivo studies, with documented solubility and storage recommendations to preserve stability.
  • Selective RORγ agonist for immunology research.
  • Orally bioavailable small-molecule modality.
  • High reported purity, useful for pharmacology studies.
  • Soluble in DMSO and ethanol; forms clear in vivo formulations at practical concentrations.
  • Stable when stored as a powder under recommended cold conditions.
  • Available in small milligram pack sizes for preclinical experiments.
Catalog No. 50-002-44514 Supplier Medchemexpress LLC Supplier No. HY10403750MG
Cintirorgon (LYC-55716) is a selective, orally bioavailable retinoic acid receptor-related orphan receptor γ (RORγ) agonist used as a research tool to modulate RORγ-dependent gene expression in T lymphocytes. It is provided as a high-purity solid suitable for in vitro and in vivo studies, with documented solubility and storage recommendations to preserve stability.
  • Selective RORγ agonist for immunology research.
  • Orally bioavailable small-molecule modality.
  • High reported purity, useful for pharmacology studies.
  • Soluble in DMSO and ethanol; forms clear in vivo formulations at practical concentrations.
  • Stable when stored as a powder under recommended cold conditions.
  • Available in small milligram pack sizes for preclinical experiments.