Learn More
Medchemexpress LLC Cintirorgon (LYC-55716) | 2055536-64-4 | MFCD31544352 | 99.1% | 603.53 g·mol⁻¹ | C27H23F6NO6S | 100 MG
SDP
Catalog No. 5000244162
Encompass_Preferred
Cintirorgon is a research-grade small molecule RORγ agonist supplied as a solid for in vitro and in vivo pharmacology. It modulates ROR signaling and is characterized by high purity, a defined molecular weight, and reported solubility in common solvents, supporting formulation and biological evaluation.
  • Selective, orally bioavailable agonist of RORγ.
  • High purity (99.1%).
  • Molecular weight 603.53 g·mol⁻¹.
  • Solid form with reported solubility: DMSO ≥113.3 mg/mL, ethanol 100 mg/mL, in vivo formulation ≥2.5 mg/mL.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Targets vitamin D related and ROR pathways.
  • Typical packaging 100 mg vial.
Catalog No. 50-002-44162 Supplier Medchemexpress LLC Supplier No. HY104037100MG
Cintirorgon is a research-grade small molecule RORγ agonist supplied as a solid for in vitro and in vivo pharmacology. It modulates ROR signaling and is characterized by high purity, a defined molecular weight, and reported solubility in common solvents, supporting formulation and biological evaluation.
  • Selective, orally bioavailable agonist of RORγ.
  • High purity (99.1%).
  • Molecular weight 603.53 g·mol⁻¹.
  • Solid form with reported solubility: DMSO ≥113.3 mg/mL, ethanol 100 mg/mL, in vivo formulation ≥2.5 mg/mL.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Targets vitamin D related and ROR pathways.
  • Typical packaging 100 mg vial.