Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS CIP 100™ Alkaline Process and Research Cleaner

Catalog No. 17463024 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
30 gal.
55 gal.
300 gal.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
17-463-024 5 gal.
50-011-72 1 gal.
17-355-272 30 gal.
17-463-004 55 gal.
17-463-022 300 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-463-024 Supplier STERIS Supplier No. 1D1005
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Specially formulated to meet the unique cleaning demands found in industries that require an effective, validatable detergent.

  • A proprietary blend of potassium hydroxide, an advanced surfactant system, and other performance-enhancing ingredients that provide multiple cleaning mechanisms
  • Removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants
  • Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
  • Ideal for use in clean-in-place (CIP), clean-out-of-place (COP), and manual applications
  • For use in pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries that require an effective, validatable detergent
  • Comprehensive support documentation package includes specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Manufactured in a cGMP compliant, FDA registered, and ISO 9001 certified facility

Specifications

Product Type Detergent
Composition A Proprietary Blend of Potassium Hydroxide (KOH)
For Use With (Application) Clean-in-Place (CIP), Clean-Out-of-Place (COP), and Manual Applications
Volume (English) 5 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.