Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ CIP 150 Alkaline Process and Research Cleaner

Catalog No. 17355268 Shop All STERIS Corporation Products
Change view
Click to view available options
Size:
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-355-268 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-355-268 Supplier STERIS™ Supplier No. 1D1505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CIP 150 Alkaline Process and Research Cleaner is a specially formulated spray-wash compound that combines an excellent alkaline detergent system with the protein cleaning properties of chlorine. It utilizes improved chemistry, significantly extending the stability of active chlorine. pH - (1%) 12.2.

CIP 150 Alkaline Process and Research Cleaner is specially formulated to meet the unique cleaning demands found in biopharmaceutical manufacturing. The blend of alkalinity, surfactants, and oxidizing agents were developed for the most challenging protein-based process residues.

HOW CIP 150 ALKALINE DETERGENT WORKS

CIP 150 Alkaline Detergent is a proprietary blend of potassium hydroxide, sodium hypochlorite (oxidizing agent), surfactants and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants, and is ideal for use in CIP, COP and manual applications.

WHY CIP 150 ALKALINE DETERGENT?

  • Flexibility - Effective cleaning against a wide variety of organic and proteinaceous process residues in clean-in-place (CIP), clean-out-of-place (COP), and manual applications.
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.

Specifications

Product Type Alkaline Cleaner
Color-coded No
Form Clear Liquid
pH 12.2 (1%)
Scent Unscented
Sterility Non-sterile
Size 5 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.