Learn More
Amsco Scientific Steris Product Number 1D2002 is STERIS’s CIP 200™ Acid Cleaner
Catalog No. NC3255062
Encompass
Product Number 1D2002 is STERIS’s CIP 200™ Acid Cleaner formulated for rouge removal and stainless steel preservation. This acid cleaner is ideal for use in CIP and COP applications.
Catalog No. NC3255062 Supplier Amsco Scientific Steris Supplier No. 1D2302
Product Number 1D2002 is STERIS’s CIP 200™ Acid Cleaner formulated for rouge removal and stainless steel preservation. This acid cleaner is ideal for use in CIP and COP applications.